Product Details

SNP ID
rs113159810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60339899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTGATGCTGGTTTCTACTGTG[C/T]TGGAGTTCTGCCTAGCTGTGCTCAC
Phenotype
MIM: 608402
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MS4A6E PubMed Links

Gene Details

Gene
MS4A6E
Gene Name
membrane spanning 4-domains A6E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139249.2 Intron NP_640342.1

View Full Product Details