Product Details

SNP ID
rs113440596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55638797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTAACACAATCATCATAGTTTGTT[A/G]TACTGGGGGATTTATACATTCTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4P4 PubMed Links

Gene Details

Gene
OR4P4
Gene Name
olfactory receptor family 4 subfamily P member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004124.2 440 Missense Mutation TAT,TGT Y147C NP_001004124.1

View Full Product Details