Product Details
- SNP ID
-
rs113798242
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:59578256 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAATGACTGGCTGTACTTTGAAACA[C/T]TCCATTTTCTCATCCCACGTCCCCA
- Phenotype
-
MIM: 167040
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OSBP
PubMed Links
Gene Details
- Gene
- OSBP
- Gene Name
- oxysterol binding protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002556.2 |
2433 |
Silent Mutation |
GAA,GAG |
E651E |
NP_002547.1 |
View Full Product Details