Product Details

SNP ID
rs114556989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73386507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCACCAGGCCTTTCTACCTGAC[C/T]GGGCAAGTCCAGTGGGGGCTCAGTC
Phenotype
MIM: 611211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RELT PubMed Links

Gene Details

Gene
RELT
Gene Name
RELT tumor necrosis factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032871.3 Intron NP_116260.2
NM_152222.1 Intron NP_689408.1
XM_005274356.2 Intron XP_005274413.1
XM_006718714.2 Intron XP_006718777.1
XM_011545305.1 Intron XP_011543607.1

View Full Product Details