Product Details

SNP ID
rs116484359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:74330539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCAGAAAAATAATTCATTCTTG[A/C]CTGCATAAATTTTACCTGGCATATT
Phenotype
MIM: 611610
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC101928580 PubMed Links

Gene Details

Gene
LOC101928580
Gene Name
uncharacterized LOC101928580
There are no transcripts associated with this gene.

Gene
PGM2L1
Gene Name
phosphoglucomutase 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173582.4 8341 UTR 3 NP_775853.2
XM_011544953.2 8341 UTR 3 XP_011543255.1

View Full Product Details