Product Details

SNP ID
rs117114945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:78172444 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCCTCTTCCAGCAGAAAATGGT[C/T]GAAAGGTATATGGAGCCACAGATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD21 PubMed Links

Gene Details

Gene
KCTD21
Gene Name
potassium channel tetramerization domain containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029859.2 2197 UTR 3 NP_001025030.1
XM_005273925.3 2197 UTR 3 XP_005273982.1
XM_005273928.1 2197 UTR 3 XP_005273985.1
XM_006718516.3 2197 UTR 3 XP_006718579.1
XM_006718517.2 2197 UTR 3 XP_006718580.1
XM_006718518.3 2197 UTR 3 XP_006718581.1
XM_011544955.2 2197 UTR 3 XP_011543257.1
Gene
KCTD21-AS1
Gene Name
KCTD21 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details