Product Details

SNP ID
rs138156775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4923608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCAGTACAGCACAGATATGAGA[A/G]ACACAGGTGTTGAGGGCTCGGAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51G1 PubMed Links

Gene Details

Gene
OR51G1
Gene Name
olfactory receptor family 51 subfamily G member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005237.1 732 Silent Mutation GTC,GTT V244V NP_001005237.1
Gene
OR51G2
Gene Name
olfactory receptor family 51 subfamily G member 2
There are no transcripts associated with this gene.

View Full Product Details