Product Details

SNP ID
rs138336362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65861979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCCAAAGCGGGAGGCTCTGG[A/C]AGCTACTGGCCAGCTCGGCACTCAG
Phenotype
MIM: 601442 MIM: 604633 MIM: 606591
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CFL1 PubMed Links

Gene Details

Gene
CFL1
Gene Name
cofilin 1
There are no transcripts associated with this gene.

Gene
EFEMP2
Gene Name
EGF containing fibulin like extracellular matrix protein 2
There are no transcripts associated with this gene.

Gene
MUS81
Gene Name
MUS81 structure-specific endonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025128.4 552 Silent Mutation GGA,GGC G128G NP_079404.3
XM_005274307.2 552 Silent Mutation GGA,GGC G128G XP_005274364.1
XM_011545269.1 552 Silent Mutation GGA,GGC G128G XP_011543571.1
XM_011545270.1 552 Silent Mutation GGA,GGC G128G XP_011543572.1
Gene
SNX32
Gene Name
sorting nexin 32
There are no transcripts associated with this gene.

View Full Product Details