Product Details

SNP ID
rs138336904
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:11964497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCCCTCCCAGCAGTGCAGCGGC[A/G]GCAGCCGCAGGCTCCCTCAGCGCCA
Phenotype
MIM: 605416 MIM: 614460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKK3 PubMed Links

Gene Details

Gene
DKK3
Gene Name
dickkopf WNT signaling pathway inhibitor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018057.1 1072 Silent Mutation GCC,GCT A340A NP_001018067.1
NM_013253.4 1072 Silent Mutation GCC,GCT A340A NP_037385.2
NM_015881.5 1072 Silent Mutation GCC,GCT A340A NP_056965.3
XM_006718178.2 1072 Silent Mutation GCC,GCT A340A XP_006718241.1
XM_017017553.1 1072 Silent Mutation GCC,GCT A354A XP_016873042.1
XM_017017554.1 1072 Silent Mutation GCC,GCT A354A XP_016873043.1
XM_017017555.1 1072 Silent Mutation GCC,GCT A354A XP_016873044.1
Gene
USP47
Gene Name
ubiquitin specific peptidase 47
There are no transcripts associated with this gene.

View Full Product Details