Product Details

SNP ID
rs138533225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6002494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCTGCACAGTATCTGAGCCTGG[A/C]AGAAAGCATGGGAACAGGAAGAGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR56A4 PubMed Links

Gene Details

Gene
OR56A4
Gene Name
olfactory receptor family 56 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005179.2 655 Missense Mutation GCC,TCC A219S NP_001005179.2

View Full Product Details