Product Details

SNP ID
rs138858028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6876830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGCACTACAAAGTCCTATGTA[C/T]TTCTTCCTCAGAAACTTGTCCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10A2 PubMed Links

Gene Details

Gene
OR10A2
Gene Name
olfactory receptor family 10 subfamily A member 2
There are no transcripts associated with this gene.

Gene
OR10A4
Gene Name
olfactory receptor family 10 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207186.2 206 Silent Mutation TAC,TAT Y61Y NP_997069.2

View Full Product Details