Product Details

SNP ID
rs138873327
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6002512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGGCAGAAAGCATGGGAACAG[A/G]AAGAGAAACAAAGGCATTCCGGGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR56A4 PubMed Links

Gene Details

Gene
OR56A4
Gene Name
olfactory receptor family 56 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005179.2 637 Missense Mutation CCT,TCT P213S NP_001005179.2

View Full Product Details