Product Details
- SNP ID
-
rs138903289
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:60337892 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATACCAACCAGACTTCTTCTTTCTT[A/C]TGATTATCATTCACCTTACACCATG
- Phenotype
-
MIM: 608402
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MS4A6E
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2289613] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MS4A6E
- Gene Name
- membrane spanning 4-domains A6E
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139249.2 |
|
Intron |
|
|
NP_640342.1 |
View Full Product Details