Product Details

SNP ID
rs138904529
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63575029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCATTCCTGCAACGCTTGCAGC[A/G]ATGATGACATCTCTGACCTGCAACA
Phenotype
MIM: 613866 MIM: 613867
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HRASLS2 PubMed Links

Gene Details

Gene
HRASLS2
Gene Name
HRAS like suppressor 2
There are no transcripts associated with this gene.

Gene
PLA2G16
Gene Name
phospholipase A2 group XVI
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128203.1 730 Silent Mutation ATC,ATT I135I NP_001121675.1
NM_007069.3 730 Silent Mutation ATC,ATT I135I NP_009000.2
XM_006718426.1 730 Silent Mutation ATC,ATT I135I XP_006718489.1
XM_011544741.1 730 Silent Mutation ATC,ATT I150I XP_011543043.1

View Full Product Details