Product Details

SNP ID
rs138952025
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75088921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCAGGTCAGCCCTGTAGGCCA[C/T]GTAGGCTATGGCAATAGATGAGTAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2AT4 PubMed Links

Gene Details

Gene
OR2AT4
Gene Name
olfactory receptor family 2 subfamily AT member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005285.1 2429 Missense Mutation ATG,GTG M265V NP_001005285.1
XM_017017655.1 2429 Missense Mutation ATG,GTG M265V XP_016873144.1
XM_017017656.1 2429 Missense Mutation ATG,GTG M265V XP_016873145.1
XM_017017657.1 2429 Missense Mutation ATG,GTG M265V XP_016873146.1
XM_017017658.1 2429 Missense Mutation ATG,GTG M265V XP_016873147.1

View Full Product Details