Product Details

SNP ID
rs139392423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123939645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCCAGCAAATCATTCCCAGGTGG[C/G]AGGATTTGTTCTACTGGGGCTCTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4D5 PubMed Links

Gene Details

Gene
OR4D5
Gene Name
olfactory receptor family 4 subfamily D member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001965.1 29 Missense Mutation GCA,GGA A10G NP_001001965.1
Gene
OR6T1
Gene Name
olfactory receptor family 6 subfamily T member 1
There are no transcripts associated with this gene.

View Full Product Details