Product Details

SNP ID
rs139413071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61876152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGAGGTGCCCGCTGAACCAGTT[A/G]GTGAAAAGTGAGGGCTCCACGTTGC
Phenotype
MIM: 606149 MIM: 606150
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FADS2 PubMed Links

Gene Details

Gene
FADS2
Gene Name
fatty acid desaturase 2
There are no transcripts associated with this gene.

Gene
FADS3
Gene Name
fatty acid desaturase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021727.4 1272 Silent Mutation ACC,ACT T373T NP_068373.1
XM_011545023.2 1272 Silent Mutation ACC,ACT T373T XP_011543325.1
XM_017017723.1 1272 Silent Mutation ACC,ACT T419T XP_016873212.1
XM_017017724.1 1272 Silent Mutation ACC,ACT T419T XP_016873213.1
Gene
MIR6746
Gene Name
microRNA 6746
There are no transcripts associated with this gene.

View Full Product Details