Product Details

SNP ID
rs139708333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60040358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCAAATAAGGAGGAAAACGAAC[A/G]CAGCTGAGCATGGGATGCCATATAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107987155 PubMed Links

Gene Details

Gene
LOC107987155
Gene Name
oocyte-secreted protein 2-like
There are no transcripts associated with this gene.

Gene
OOSP2
Gene Name
oocyte secreted protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173801.4 84 UTR 5 NP_776162.2

View Full Product Details