Product Details
- SNP ID
-
rs139734163
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:8094145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAAGGCGGCAGCTACAGCAGGGG[A/G]CCAGGGTGGCGCCGCTAGGAAGGAG
- Phenotype
-
MIM: 610509
MIM: 601197
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RIC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4758289] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RIC3
- Gene Name
- RIC3 acetylcholine receptor chaperone
- Gene
- TUB
- Gene Name
- tubby bipartite transcription factor
View Full Product Details