Product Details
- SNP ID
-
rs139771453
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:61339157 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAGCATCCTGGCAGCCATCAGGGC[C/T]GTGGCCCAGGCCGGCACAGGTAAGC
- Phenotype
-
MIM: 600045
MIM: 615844
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYB561A3
PubMed Links
Gene Details
- Gene
- CYB561A3
- Gene Name
- cytochrome b561 family member A3
There are no transcripts associated with this gene.
- Gene
- DDB1
- Gene Name
- damage specific DNA binding protein 1
There are no transcripts associated with this gene.
- Gene
- TKFC
- Gene Name
- triokinase and FMN cyclase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015533.3 |
672 |
Silent Mutation |
GCC,GCT |
A95A |
NP_056348.2 |
XM_011544912.2 |
672 |
Silent Mutation |
GCC,GCT |
A141A |
XP_011543214.1 |
XM_017017517.1 |
672 |
Silent Mutation |
GCC,GCT |
A141A |
XP_016873006.1 |
XM_017017518.1 |
672 |
Silent Mutation |
GCC,GCT |
A141A |
XP_016873007.1 |
XM_017017519.1 |
672 |
Silent Mutation |
GCC,GCT |
A95A |
XP_016873008.1 |
XM_017017520.1 |
672 |
Silent Mutation |
GCC,GCT |
A141A |
XP_016873009.1 |
XM_017017521.1 |
672 |
Silent Mutation |
GCC,GCT |
A95A |
XP_016873010.1 |
XM_017017522.1 |
672 |
Silent Mutation |
GCC,GCT |
A95A |
XP_016873011.1 |
XM_017017523.1 |
672 |
Silent Mutation |
GCC,GCT |
A95A |
XP_016873012.1 |
XM_017017524.1 |
672 |
Silent Mutation |
GCC,GCT |
A25A |
XP_016873013.1 |
View Full Product Details