Product Details

SNP ID
rs140020543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67365423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGCAGAAGTGGGCCAGGCTGC[A/G]GCGCAGCTCAGCAGTGGCAGCCTGA
Phenotype
MIM: 607672
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLCF1 PubMed Links

Gene Details

Gene
CLCF1
Gene Name
cardiotrophin-like cytokine factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166212.1 506 Missense Mutation CGC,TGC R121C NP_001159684.1
NM_013246.2 506 Missense Mutation CGC,TGC R131C NP_037378.1
Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

View Full Product Details