Product Details

SNP ID
rs140450102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111882598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCCCAAACCCTTGCCTTCCCA[A/G]GTAATCTAACTTTGTCTTTTTTGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf1 PubMed Links

Gene Details

Gene
C11orf1
Gene Name
chromosome 11 open reading frame 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022761.2 449 Silent Mutation CAA,CAG Q92Q NP_073598.1
XM_011542957.2 449 Silent Mutation CAA,CAG Q108Q XP_011541259.1
XM_011542958.2 449 Silent Mutation CAA,CAG Q108Q XP_011541260.1
XM_017018129.1 449 Silent Mutation CAA,CAG Q92Q XP_016873618.1
Gene
FDXACB1
Gene Name
ferredoxin-fold anticodon binding domain containing 1
There are no transcripts associated with this gene.

View Full Product Details