Product Details

SNP ID
rs140479762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:126304214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCTCCTGTCCGCTTACCGTTCT[C/G]CGGCTTCAGACTGCAGAAGGTGCTG
Phenotype
MIM: 610534 MIM: 606252
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCPS PubMed Links

Gene Details

Gene
DCPS
Gene Name
decapping enzyme, scavenger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014026.4 914 Missense Mutation TCC,TGC S45C NP_054745.1
XM_011542778.2 914 Missense Mutation TCC,TGC S45C XP_011541080.1
XM_011542780.2 914 Intron XP_011541082.1
XM_017017612.1 914 UTR 5 XP_016873101.1
Gene
HP11014
Gene Name
uncharacterized HP11014
There are no transcripts associated with this gene.

Gene
TIRAP
Gene Name
TIR domain containing adaptor protein
There are no transcripts associated with this gene.

View Full Product Details