Product Details
- SNP ID
-
rs140492814
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:114439660 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGAGCATGGCTCAGCGGATGGT[C/T]TGGGTGGACCTGGAGGTGAGTGAGG
- Phenotype
-
MIM: 607149
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
REXO2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs476200] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- REXO2
- Gene Name
- RNA exonuclease 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015523.3 |
275 |
Silent Mutation |
GTC,GTT |
V44V |
NP_056338.2 |
View Full Product Details