Product Details

SNP ID
rs140566640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58267374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAGAAGTGCTCAATGCCCTGA[G/T]CCTGGCAGAATGGCAGAGAGAAGAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10W1 PubMed Links

Gene Details

Gene
OR10W1
Gene Name
olfactory receptor family 10 subfamily W member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207374.3 887 Missense Mutation NP_997257.2

View Full Product Details