Product Details

SNP ID
rs140670344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116844109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGTATTCAAGGTCTCTCTCTTGT[G/T]TTGAAGATCAGAAGGAGATAACCCA
Phenotype
MIM: 107680 MIM: 614776
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APOA1 PubMed Links

Gene Details

Gene
APOA1
Gene Name
apolipoprotein A1
There are no transcripts associated with this gene.

Gene
APOA1-AS
Gene Name
APOA1 antisense RNA
There are no transcripts associated with this gene.

Gene
SIK3
Gene Name
SIK family kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281748.1 5358 UTR 3 NP_001268677.1
NM_001281749.1 5358 UTR 3 NP_001268678.1
NM_025164.4 5358 UTR 3 NP_079440.3
XM_005271481.2 5358 UTR 3 XP_005271538.1
XM_005271482.4 5358 UTR 3 XP_005271539.2
XM_005271484.3 5358 UTR 3 XP_005271541.2
XM_005271485.3 5358 UTR 3 XP_005271542.1
XM_011542721.2 5358 Intron XP_011541023.1
XM_011542722.1 5358 UTR 3 XP_011541024.1
XM_011542723.2 5358 UTR 3 XP_011541025.1
XM_011542724.2 5358 UTR 3 XP_011541026.1
XM_011542725.2 5358 UTR 3 XP_011541027.1
XM_011542726.1 5358 Intron XP_011541028.1
XM_017017424.1 5358 UTR 3 XP_016872913.1
XM_017017425.1 5358 UTR 3 XP_016872914.1
XM_017017426.1 5358 UTR 3 XP_016872915.1
XM_017017427.1 5358 UTR 3 XP_016872916.1

View Full Product Details