Product Details

SNP ID
rs140788036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62735447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGAATATAGTCAGAAAGTCCTG[C/G]AACGACAGCCTGATAATGCCAAGGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HNRNPUL2 PubMed Links

Gene Details

Gene
HNRNPUL2
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
There are no transcripts associated with this gene.

Gene
HNRNPUL2-BSCL2
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TTC9C
Gene Name
tetratricopeptide repeat domain 9C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318812.1 430 Missense Mutation CAA,GAA Q102E NP_001305741.1
NM_001318813.1 430 Missense Mutation CAA,GAA Q102E NP_001305742.1
NM_001318814.1 430 Missense Mutation CAA,GAA Q18E NP_001305743.1
NM_001318815.1 430 Missense Mutation CAA,GAA Q18E NP_001305744.1
NM_001318816.1 430 Missense Mutation CAA,GAA Q18E NP_001305745.1
NM_173810.3 430 Missense Mutation CAA,GAA Q102E NP_776171.1

View Full Product Details