Product Details

SNP ID
rs140822122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57551671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTGCACTCCAGAATTCATCTCT[A/G]ACTTTAATGGCTTAAGCAAGAACAT
Phenotype
MIM: 613664 MIM: 603890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMTNL1 PubMed Links

Gene Details

Gene
SMTNL1
Gene Name
smoothelin like 1
There are no transcripts associated with this gene.

Gene
UBE2L6
Gene Name
ubiquitin conjugating enzyme E2 L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004223.4 1245 UTR 3 NP_004214.1
NM_198183.2 1245 UTR 3 NP_937826.1

View Full Product Details