Product Details

SNP ID
rs141029883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61368635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTACTTCTATAAACGGACAGCC[A/G]TAAGACTAGGCGATCCTCACTTCTA
Phenotype
MIM: 614459
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYB561A3 PubMed Links

Gene Details

Gene
CYB561A3
Gene Name
cytochrome b561 family member A3
There are no transcripts associated with this gene.

Gene
TMEM138
Gene Name
transmembrane protein 138
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016464.4 469 Missense Mutation ATA,GTA I139V NP_057548.1
XM_006718585.3 469 UTR 3 XP_006718648.1
XM_006718586.2 469 Missense Mutation ATA,GTA I138V XP_006718649.1
XM_006718588.3 469 Missense Mutation ATA,GTA I81V XP_006718651.1
XM_011545098.2 469 Intron XP_011543400.1
XM_011545099.1 469 Intron XP_011543401.1
XM_011545100.1 469 Intron XP_011543402.1
XM_017017917.1 469 Intron XP_016873406.1

View Full Product Details