Product Details

SNP ID
rs141055431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72579002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGTGCTGCTTGTTGTTTCGGTC[A/G]TAGCCCACTGTTGAGGGGAGGATGG
Phenotype
MIM: 602658
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01537 PubMed Links

Gene Details

Gene
LINC01537
Gene Name
long intergenic non-protein coding RNA 1537
There are no transcripts associated with this gene.

Gene
PDE2A
Gene Name
phosphodiesterase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143839.3 2843 Silent Mutation TAC,TAT Y781Y NP_001137311.1
NM_001146209.2 2843 Silent Mutation TAC,TAT Y779Y NP_001139681.1
NM_001243784.1 2843 Silent Mutation TAC,TAT Y767Y NP_001230713.1
NM_002599.4 2843 Silent Mutation TAC,TAT Y788Y NP_002590.1
XM_005274040.2 2843 Silent Mutation TAC,TAT Y787Y XP_005274097.1
XM_011545096.2 2843 Silent Mutation TAC,TAT Y467Y XP_011543398.1
XM_017017910.1 2843 Silent Mutation TAC,TAT Y467Y XP_016873399.1

View Full Product Details