Product Details

SNP ID
rs141381613
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5884328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGACACCCACTTCTATCCCCC[A/C]TTCTTCCTCCTGCTAGGAATACCAG
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
OR52E4 PubMed Links

Gene Details

Gene
OR52E4
Gene Name
olfactory receptor family 52 subfamily E member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005165.1 36 Silent Mutation CCA,CCC P12P NP_001005165.1

View Full Product Details