Product Details
- SNP ID
-
rs141586285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5059066 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTGATGCTGGCACAAGATAGAT[A/G]AGCAAGACCCATGTGCTCACAGTAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52E2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11035396,rs2445333] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52E2
- Gene Name
- olfactory receptor family 52 subfamily E member 2
View Full Product Details