Product Details

SNP ID
rs142291500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34882782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTCCTACTTTCTTCATTGATA[A/C]GGCAATATCAAATAAATAGTCATAA
Phenotype
MIM: 612491
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APIP PubMed Links

Gene Details

Gene
APIP
Gene Name
APAF1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015957.3 791 Missense Mutation ATA,GTA I222V NP_057041.2
XM_011520154.2 791 Intron XP_011518456.1
XM_017017875.1 791 Intron XP_016873364.1

View Full Product Details