Product Details

SNP ID
rs142320326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:644595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACGCTTTCAGCCACGTGGACTTC[A/G]TCTGCCTGGACGGTGACAGCTGCTG
Phenotype
MIM: 602635 MIM: 126452
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DEAF1 PubMed Links

Gene Details

Gene
DEAF1
Gene Name
DEAF1, transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293634.1 2360 Silent Mutation GAC,GAT D476D NP_001280563.1
NM_021008.3 2360 Silent Mutation GAC,GAT D551D NP_066288.2
XM_011519842.2 2360 Intron XP_011518144.1
Gene
DRD4
Gene Name
dopamine receptor D4
There are no transcripts associated with this gene.

View Full Product Details