Product Details

SNP ID
rs142444776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:75089671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGGATGCCCAATAGATAGAAGA[C/T]GGGTGAGCCATCCACTGATTCATTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2AT4 PubMed Links

Gene Details

Gene
OR2AT4
Gene Name
olfactory receptor family 2 subfamily AT member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005285.1 1679 Missense Mutation ATC,GTC I15V NP_001005285.1
XM_017017655.1 1679 Missense Mutation ATC,GTC I15V XP_016873144.1
XM_017017656.1 1679 Missense Mutation ATC,GTC I15V XP_016873145.1
XM_017017657.1 1679 Missense Mutation ATC,GTC I15V XP_016873146.1
XM_017017658.1 1679 Missense Mutation ATC,GTC I15V XP_016873147.1

View Full Product Details