Product Details

SNP ID
rs142656457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5856790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAAATCCTCAGCACTCTCTCTC[A/G]AATCTGCTTTGTCCTGACTCCATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52E8 PubMed Links

Gene Details

Gene
OR52E8
Gene Name
olfactory receptor family 52 subfamily E member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005168.1 913 Nonsense Mutation CGA,TGA R305* NP_001005168.1

View Full Product Details