Product Details

SNP ID
rs142814072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55343331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATGGTGGGAAACCTCCTCATTT[A/G]GGTGACTACTATTGGCAGCCCCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4A16 PubMed Links

Gene Details

Gene
OR4A16
Gene Name
olfactory receptor family 4 subfamily A member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005274.1 131 Missense Mutation TAG,TGG *44W NP_001005274.1

View Full Product Details