Product Details

SNP ID
rs142862260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10514925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTTCACTTCAAGGTCATGGCCT[A/G]TGGGGCTGGCCTGCCTCATCAGCCA
Phenotype
MIM: 102772 MIM: 616641
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMPD3 PubMed Links

Gene Details

Gene
AMPD3
Gene Name
adenosine monophosphate deaminase 3
There are no transcripts associated with this gene.

Gene
MIR4485
Gene Name
microRNA 4485
There are no transcripts associated with this gene.

Gene
MTRNR2L8
Gene Name
MT-RNR2-like 8
There are no transcripts associated with this gene.

Gene
RNF141
Gene Name
ring finger protein 141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016422.3 837 Silent Mutation CAC,CAT H228H NP_057506.2

View Full Product Details