Product Details
- SNP ID
-
rs143094813
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:64341644 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGGTGCTGGCACTGTCTGGGCCA[G/T]ATCCTGGGGAGCTGGCACCTGCCGA
- Phenotype
-
MIM: 611205
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC88B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs647152] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC88B
- Gene Name
- coiled-coil domain containing 88B
- Gene
- MIR7155
- Gene Name
- microRNA 7155
There are no transcripts associated with this gene.
View Full Product Details