Product Details

SNP ID
rs143133987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3655028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAAATCTCAGTGGCAAACAAAC[A/G]TTTACTCCCAGGCTCATGGTCTTTG
Phenotype
MIM: 601625
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ART1 PubMed Links

Gene Details

Gene
ART1
Gene Name
ADP-ribosyltransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004314.2 Intron NP_004305.2
XM_011520114.2 Intron XP_011518416.1
XM_017017763.1 Intron XP_016873252.1

View Full Product Details