Product Details

SNP ID
rs143204032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:89715169 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCTCTTTATAGGATTTGGGAGA[C/T]GTAATGAAAAGGTAAGTTTGCCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM77 PubMed Links

Gene Details

Gene
TRIM77
Gene Name
tripartite motif containing 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146162.1 Intron NP_001139634.1
NM_001271942.1 Intron NP_001258871.1

View Full Product Details