Product Details

SNP ID
rs143300353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72814491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCCCCCGCAGCGCGCTGGAAA[C/T]GCCACATCGTGCGGCAGCTGCGGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG16L2 PubMed Links

Gene Details

Gene
ATG16L2
Gene Name
autophagy related 16 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318766.1 65 UTR 5 NP_001305695.1
NM_033388.1 65 Missense Mutation CGC,TGC R16C NP_203746.1
XM_005274376.4 65 Missense Mutation CGC,TGC R16C XP_005274433.1
XM_006718732.2 65 Missense Mutation CGC,TGC R16C XP_006718795.1
XM_006718733.3 65 Missense Mutation CGC,TGC R16C XP_006718796.1
XM_006718734.2 65 Intron XP_006718797.1
XM_011545332.1 65 Missense Mutation CGC,TGC R16C XP_011543634.1
XM_011545333.1 65 UTR 5 XP_011543635.1
XM_011545334.1 65 Intron XP_011543636.1

View Full Product Details