Product Details

SNP ID
rs143338269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55638716 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTCACAGGGATGGCCTATGACC[A/G]CTATGTGGCCATTTGCAAGCCCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4P4 PubMed Links

Gene Details

Gene
OR4P4
Gene Name
olfactory receptor family 4 subfamily P member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004124.2 359 Missense Mutation CAC,CGC H120R NP_001004124.1

View Full Product Details