Product Details

SNP ID
rs143538624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6717389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGCCCAAAAAGGGCATTCAGCAC[A/G]GTGGATTTCCCTGAGCTCTGCAAGC
Phenotype
MIM: 616121
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GVINP1 PubMed Links

Gene Details

Gene
GVINP1
Gene Name
GTPase, very large interferon inducible pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details