Product Details

SNP ID
rs143633691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:105041463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAACCTGGGATTCTAGGATATAG[A/T]GATCCTTTATGCAAGGGGAGCAGAA
Phenotype
MIM: 615680 MIM: 147678
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CARD16 PubMed Links

Gene Details

Gene
CARD16
Gene Name
caspase recruitment domain family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017534.1 621 Silent Mutation TCA,TCT S177S NP_001017534.1
NM_052889.2 621 UTR 3 NP_443121.1
XM_011542583.2 621 Silent Mutation TCA,TCT S245S XP_011540885.1
Gene
CASP1
Gene Name
caspase 1
There are no transcripts associated with this gene.

View Full Product Details