Product Details

SNP ID
rs144016594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:95067617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGGAGCCGCAGCCCCAGGCGG[A/C]GACACCGCAGCCGATCCCGGGGTCC
Phenotype
MIM: 603269
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SRSF8 PubMed Links

Gene Details

Gene
SRSF8
Gene Name
serine and arginine rich splicing factor 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032102.3 741 Silent Mutation AGA,CGA R131R NP_115285.1
XM_017017142.1 741 Silent Mutation AGA,CGA R131R XP_016872631.1

View Full Product Details