Product Details
- SNP ID
-
rs144141685
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:13389032 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGTCTACATGCCGACTCTTTCC[C/T]TCTTCCTTCTCCCAGGACATTCGAA
- Phenotype
-
MIM: 602550
MIM: 615933
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARNTL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3833472] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARNTL
- Gene Name
- aryl hydrocarbon receptor nuclear translocator like
There are no transcripts associated with this gene.
- Gene
- BTBD10
- Gene Name
- BTB domain containing 10
View Full Product Details