Product Details
- SNP ID
-
rs144383299
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:60043606 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATCGGATACATACATATGTATAT[A/G]AGTTTATATATCTTGTTCGTGATTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OOSP2
PubMed Links
Gene Details
- Gene
- OOSP2
- Gene Name
- oocyte secreted protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173801.4 |
387 |
Missense Mutation |
AAG,GAG |
K68E |
NP_776162.2 |
View Full Product Details