Product Details
- SNP ID
-
rs144458899
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:134256589 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAAGAGCAGAAAGAATTTCAAAAA[A/G]TGGCCTTTGACTTTGCTGCCCGAGA
- Phenotype
-
MIM: 604773
MIM: 613739
MIM: 610027
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ACAD8
PubMed Links
Gene Details
- Gene
- ACAD8
- Gene Name
- acyl-CoA dehydrogenase family member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014384.2 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
NP_055199.1 |
XM_005271505.3 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_005271562.1 |
XM_011542750.2 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_011541052.1 |
XM_017017542.1 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873031.1 |
XM_017017543.1 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873032.1 |
XM_017017544.1 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873033.1 |
XM_017017545.1 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873034.1 |
XM_017017546.1 |
220 |
Silent Mutation |
AAA,AAG |
K9K |
XP_016873035.1 |
XM_017017547.1 |
220 |
Silent Mutation |
AAA,AAG |
K9K |
XP_016873036.1 |
XM_017017548.1 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873037.1 |
XM_017017549.1 |
220 |
Missense Mutation |
ATG,GTG |
M51V |
XP_016873038.1 |
- Gene
- THYN1
- Gene Name
- thymocyte nuclear protein 1
There are no transcripts associated with this gene.
- Gene
- VPS26B
- Gene Name
- VPS26, retromer complex component B
There are no transcripts associated with this gene.
View Full Product Details