Product Details

SNP ID
rs144508173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64340630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGCTGGGACTGGCCGGGCTGGT[C/T]GGGGAGGCGGAGGACTCGGAGGGGG
Phenotype
MIM: 611205
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC88B PubMed Links

Gene Details

Gene
CCDC88B
Gene Name
coiled-coil domain containing 88B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032251.5 146 Silent Mutation GTC,GTT V28V NP_115627.6
XM_006718519.3 146 Silent Mutation GTC,GTT V28V XP_006718582.1
XM_011544957.2 146 Silent Mutation GTC,GTT V28V XP_011543259.1
XM_011544959.2 146 Silent Mutation GTC,GTT V28V XP_011543261.1
XM_011544960.2 146 Silent Mutation GTC,GTT V28V XP_011543262.1
XM_011544962.2 146 Intron XP_011543264.1
XM_017017601.1 146 Silent Mutation GTC,GTT V28V XP_016873090.1
Gene
MIR7155
Gene Name
microRNA 7155
There are no transcripts associated with this gene.

View Full Product Details